The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016224.4 | NP_057308.1 | 16 - 17 | 1907 | 95 |
XM_005267015.2 | XP_005267072.1 | 18 - 19 | 1985 | 95 | |
XM_011535886.2 | XP_011534188.1 | 13 - 14 | 1446 | 95 | |
GenBank mRNA | AF076957.1 | - | 11 - 12 | 1127 | 95 |
AF121859.1 | - | 16 - 17 | 1667 | 95 | |
AF131214.1 | - | 16 - 17 | 1686 | 95 | |
AF172847.1 | - | 16 - 17 | 1787 | 95 | |
AK026637.1 | - | 13 - 14 | 1452 | 95 | |
AK127345.1 | - | 13 - 14 | 1762 | 95 | |
AK314360.1 | - | 16 - 17 | 1741 | 95 | |
BC001084.2 | - | 7 - 8 | 708 | 95 | |
BC005022.1 | - | 16 - 17 | 1814 | 95 |