The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016224.4 | NP_057308.1 | 17 - 18 | 2000 | 60 |
XM_005267015.2 | XP_005267072.1 | 19 - 20 | 2078 | 60 | |
XM_011535886.2 | XP_011534188.1 | 14 - 15 | 1539 | 60 | |
GenBank mRNA | AB209736.1 | - | 10 - 11 | 5968 | 60 |
AF076957.1 | - | 12 - 13 | 1220 | 60 | |
AF121859.1 | - | 17 - 18 | 1760 | 60 | |
AF131214.1 | - | 17 - 18 | 1779 | 60 | |
AF172847.1 | - | 17 - 18 | 1880 | 60 | |
AK026637.1 | - | 14 - 15 | 1545 | 60 | |
AK127345.1 | - | 14 - 15 | 1855 | 60 | |
AK314360.1 | - | 17 - 18 | 1834 | 60 | |
BC001084.2 | - | 8 - 9 | 801 | 60 | |
BC005022.1 | - | 17 - 18 | 1907 | 60 |