The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016224.4 | NP_057308.1 | 9 - 10 | 1218 | 75 |
XM_005267015.2 | XP_005267072.1 | 11 - 12 | 1296 | 75 | |
XM_011535886.2 | XP_011534188.1 | 6 - 7 | 757 | 75 | |
GenBank mRNA | AB209736.1 | - | 5 - 6 | 549 | 75 |
AF076957.1 | - | 4 - 5 | 438 | 75 | |
AF121859.1 | - | 9 - 10 | 978 | 75 | |
AF131214.1 | - | 9 - 10 | 997 | 75 | |
AF172847.1 | - | 9 - 10 | 1098 | 75 | |
AK026637.1 | - | 6 - 7 | 763 | 75 | |
AK127345.1 | - | 6 - 7 | 1073 | 75 | |
AK314360.1 | - | 9 - 10 | 1052 | 75 | |
BC005022.1 | - | 9 - 10 | 1125 | 75 |