Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
Chromosome Location
Chr.9: 14734666 - 14911653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144966.5 NP_659403.4 4 - 5 1139 142
XM_005251382.3 XP_005251439.1 3 - 4 1387 142
XM_017014316.1 XP_016869805.1 3 - 4 1387 142
XM_017014317.1 XP_016869806.1 4 - 5 930 142
XM_017014318.1 XP_016869807.1 4 - 5 1685 142
XM_017014319.1 XP_016869808.1 5 - 6 1589 142
XM_017014320.1 XP_016869809.1 4 - 5 1516 142
XM_017014321.1 XP_016869810.1 3 - 4 1179 142
XM_017014322.1 XP_016869811.1 3 - 4 973 142
XM_017014323.1 XP_016869812.1 3 - 4 646 142
XM_017014324.1 XP_016869813.1 3 - 4 1387 142
XM_017014325.1 XP_016869814.1 3 - 4 1387 142
XM_017014326.1 XP_016869815.1 2 - 3 203 142
XM_017014328.1 XP_016869817.1 3 - 4 1387 142
XM_017014329.1 XP_016869818.1 3 - 4 1387 142
XM_017014330.1 XP_016869819.1 3 - 4 1387 142
GenBank mRNA AB160987.1 - 2 - 3 323 142
BX537574.1 - 4 - 5 1140 142

View Full Product Details