The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000806.5 | NP_000797.2 | 10 - 11 | 1531 | 85 |
NM_001127643.1 | NP_001121115.1 | 10 - 11 | 1418 | 85 | |
NM_001127644.1 | NP_001121116.1 | 9 - 10 | 1567 | 85 | |
NM_001127645.1 | NP_001121117.1 | 9 - 10 | 1155 | 85 | |
NM_001127648.1 | NP_001121120.1 | 8 - 9 | 1177 | 85 | |
GenBank mRNA | AK223447.1 | - | 9 - 10 | 1304 | 85 |
BC030696.1 | - | 9 - 10 | 1132 | 85 | |
X14766.1 | - | 9 - 10 | 1277 | 85 |