The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004262.2 | NP_004253.1 | 6 - 7 | 578 | 83 |
XM_005265710.4 | XP_005265767.1 | 5 - 6 | 1521 | 83 | |
GenBank mRNA | AB002134.1 | - | 6 - 7 | 574 | 83 |
AK290289.1 | - | 6 - 7 | 579 | 83 | |
AK296853.1 | - | 5 - 6 | 441 | 83 | |
BC125195.1 | - | 6 - 7 | 563 | 83 | |
BC125196.1 | - | 6 - 7 | 563 | 83 |