The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010985.2 | NP_001010985.2 | 5 - 6 | 779 | 56 |
NM_001265613.1 | NP_001252542.1 | 5 - 6 | 710 | 56 | |
XM_017001173.1 | XP_016856662.1 | 5 - 6 | 3132 | 56 | |
XM_017001174.1 | XP_016856663.1 | 5 - 6 | 3132 | 56 | |
XM_017001175.1 | XP_016856664.1 | 5 - 6 | 3063 | 56 | |
GenBank mRNA | AK129834.1 | - | 5 - 6 | 779 | 56 |
BC132900.1 | - | 5 - 6 | 756 | 56 | |
BC132902.1 | - | 5 - 6 | 756 | 56 | |
BC144471.1 | - | 5 - 6 | 687 | 56 |