Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THEG
Entrez Gene ID
51298
Gene Name
theg spermatid protein
Gene Aliases
CT56, THEG1
Chromosome Location
Chr.19: 362057 - 376693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016585.4 NP_057669.1 2 - 3 479 66
XM_011528049.2 XP_011526351.1 2 - 3 1158 66
XM_011528052.2 XP_011526354.1 2 - 3 1157 66
GenBank mRNA AB033129.1 - 2 - 3 479 66
AF268610.1 - 2 - 3 445 66
BC028574.2 - 2 - 3 458 66

View Full Product Details