The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016585.4 | NP_057669.1 | 3 - 4 | 552 | 61 |
XM_011528049.2 | XP_011526351.1 | 3 - 4 | 1231 | 61 | |
XM_011528052.2 | XP_011526354.1 | 3 - 4 | 1230 | 61 | |
GenBank mRNA | AB033129.1 | - | 3 - 4 | 552 | 61 |
AF268610.1 | - | 3 - 4 | 518 | 61 | |
BC028574.2 | - | 3 - 4 | 531 | 61 |