The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032951.2 | NP_116569.1 | 14 - 15 | 2236 | 98 |
NM_032952.2 | NP_116570.1 | 14 - 15 | 2179 | 98 | |
NM_032953.2 | NP_116571.1 | 14 - 15 | 2230 | 98 | |
NM_032954.2 | NP_116572.1 | 14 - 15 | 2173 | 98 | |
NR_134541.1 | 13 - 14 | 2088 | 98 | ||
XM_011516277.1 | XP_011514579.1 | 14 - 15 | 2413 | 98 | |
XM_011516278.1 | XP_011514580.1 | 14 - 15 | 2410 | 98 | |
XM_011516279.1 | XP_011514581.1 | 14 - 15 | 2359 | 98 | |
XM_011516281.2 | XP_011514583.1 | 8 - 9 | 1910 | 98 | |
XM_017012263.1 | XP_016867752.1 | 9 - 10 | 1432 | 98 | |
GenBank mRNA | AF156603.1 | - | 14 - 15 | 2236 | 98 |
AF245470.1 | - | 14 - 15 | 2188 | 98 | |
AF245471.1 | - | 14 - 15 | 2131 | 98 | |
AF245472.1 | - | 14 - 15 | 2182 | 98 | |
AF245473.1 | - | 14 - 15 | 2125 | 98 | |
AF245474.1 | - | 13 - 14 | 2037 | 98 | |
BC012925.1 | - | 13 - 14 | 1928 | 98 |