Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYH11
Entrez Gene ID
4629
Gene Name
myosin heavy chain 11
Gene Aliases
AAT4, FAA4, SMHC, SMMHC
Chromosome Location
Chr.16: 15703135 - 15857032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040113.1 NP_001035202.1 1 - 2 91 72
NM_001040114.1 NP_001035203.1 1 - 2 91 72
NM_002474.2 NP_002465.1 1 - 2 91 72
NM_022844.2 NP_074035.1 1 - 2 91 72
XM_011522502.2 XP_011520804.1 1 - 2 93 72
XM_017023250.1 XP_016878739.1 1 - 2 55 72
GenBank mRNA AB020673.2 - 1 - 2 59 72
AK308389.1 - 1 - 2 88 72
AY520816.1 - 1 - 2 72 72
AY520817.1 - 1 - 2 72 72
BC031040.1 - 1 - 2 89 72
BC101677.1 - 1 - 2 55 72
BC104906.1 - 1 - 2 55 72
BC143364.1 - 1 - 2 55 72
BX489788.1 - 1 - 2 91 72
EU489063.1 - 1 - 2 91 72

View Full Product Details