The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005399.4 | NP_005390.1 | 6 - 7 | 859 | 94 |
NR_103870.1 | 4 - 5 | 571 | 94 | ||
NR_103871.1 | 5 - 6 | 692 | 94 | ||
XM_011509729.1 | XP_011508031.1 | 6 - 7 | 859 | 94 | |
XM_017001697.1 | XP_016857186.1 | 5 - 6 | 738 | 94 | |
GenBank mRNA | AJ224538.1 | - | 5 - 6 | 675 | 94 |
AK292820.1 | - | 6 - 7 | 740 | 94 | |
AK294863.1 | - | 5 - 6 | 664 | 94 | |
AK304069.1 | - | 4 - 5 | 452 | 94 | |
AK310982.1 | - | 6 - 7 | 740 | 94 | |
AK316005.1 | - | 5 - 6 | 570 | 94 | |
AK316183.1 | - | 4 - 5 | 543 | 94 | |
BC053610.1 | - | 6 - 7 | 814 | 94 | |
BU166852.1 | - | 2 - 3 | 176 | 94 | |
KJ534919.1 | - | 5 - 6 | 675 | 94 | |
KJ535024.1 | - | 4 - 5 | 508 | 94 | |
KJ535025.1 | - | 5 - 6 | 675 | 94 |