Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTK
Entrez Gene ID
695
Gene Name
Bruton tyrosine kinase
Gene Aliases
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
Chromosome Location
Chr.X: 101349447 - 101390796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000061.2 NP_000052.1 2 - 3 327 158
NM_001287344.1 NP_001274273.1 2 - 3 483 158
NM_001287345.1 NP_001274274.1 3 - 4 492 158
GenBank mRNA AK057105.1 - 3 - 4 446 158
AK225577.1 - 2 - 3 289 158
AK309851.1 - 2 - 3 469 158
AK314382.1 - 2 - 3 294 158
BC109079.1 - 2 - 3 259 158
BC109080.1 - 2 - 3 259 158
DA619542.1 - 2 - 3 327 158
KF241986.1 - 1 - 2 134 158
X58957.1 - 2 - 3 266 158

View Full Product Details