Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAP2
Entrez Gene ID
23527
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Gene Aliases
CENTB2, CNT-B2
Chromosome Location
Chr.3: 195274736 - 195443176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012287.5 NP_036419.3 18 - 19 2000 105
XM_006713557.3 XP_006713620.1 19 - 20 2021 105
XM_011512602.2 XP_011510904.1 20 - 21 2126 105
XM_011512603.2 XP_011510905.1 19 - 20 2105 105
XM_011512604.2 XP_011510906.1 21 - 22 2285 105
XM_011512605.2 XP_011510907.1 20 - 21 2047 105
XM_011512606.2 XP_011510908.1 20 - 21 2028 105
XM_017006046.1 XP_016861535.1 21 - 22 2234 105
XM_017006047.1 XP_016861536.1 20 - 21 2213 105
XM_017006048.1 XP_016861537.1 20 - 21 2129 105
XM_017006049.1 XP_016861538.1 19 - 20 2108 105
GenBank mRNA AJ238248.1 - 18 - 19 1819 105
AK290369.1 - 18 - 19 1932 105
BC031005.1 - 1 - 2 28 105
BC060767.1 - 18 - 19 1935 105
D26069.3 - 18 - 19 1769 105

View Full Product Details