The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012287.5 | NP_036419.3 | 4 - 5 | 524 | 101 |
XM_006713557.3 | XP_006713620.1 | 4 - 5 | 524 | 101 | |
XM_011512602.2 | XP_011510904.1 | 4 - 5 | 524 | 101 | |
XM_011512603.2 | XP_011510905.1 | 4 - 5 | 524 | 101 | |
XM_011512604.2 | XP_011510906.1 | 5 - 6 | 683 | 101 | |
XM_011512605.2 | XP_011510907.1 | 4 - 5 | 445 | 101 | |
XM_011512606.2 | XP_011510908.1 | 4 - 5 | 426 | 101 | |
GenBank mRNA | AK222635.1 | - | 4 - 5 | 479 | 101 |
AK290369.1 | - | 4 - 5 | 456 | 101 | |
BC060767.1 | - | 4 - 5 | 459 | 101 | |
D26069.3 | - | 4 - 5 | 293 | 101 | |
DA774230.1 | - | 4 - 5 | 479 | 101 | |
DB087306.1 | - | 4 - 5 | 524 | 101 |