The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012287.5 | NP_036419.3 | 7 - 8 | 805 | 96 |
XM_006713557.3 | XP_006713620.1 | 7 - 8 | 805 | 96 | |
XM_011512602.2 | XP_011510904.1 | 7 - 8 | 805 | 96 | |
XM_011512603.2 | XP_011510905.1 | 7 - 8 | 805 | 96 | |
XM_011512604.2 | XP_011510906.1 | 8 - 9 | 964 | 96 | |
XM_011512605.2 | XP_011510907.1 | 7 - 8 | 726 | 96 | |
XM_011512606.2 | XP_011510908.1 | 7 - 8 | 707 | 96 | |
XM_017006046.1 | XP_016861535.1 | 8 - 9 | 913 | 96 | |
XM_017006047.1 | XP_016861536.1 | 8 - 9 | 913 | 96 | |
XM_017006048.1 | XP_016861537.1 | 8 - 9 | 913 | 96 | |
XM_017006049.1 | XP_016861538.1 | 8 - 9 | 913 | 96 | |
GenBank mRNA | AJ238248.1 | - | 7 - 8 | 624 | 96 |
AK290369.1 | - | 7 - 8 | 737 | 96 | |
BC060767.1 | - | 7 - 8 | 740 | 96 | |
D26069.3 | - | 7 - 8 | 574 | 96 |