The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321463.1 | NP_001308392.1 | 5 - 6 | 854 | 64 |
NM_020170.3 | NP_064555.2 | 5 - 6 | 854 | 64 | |
GenBank mRNA | AK125002.1 | - | 5 - 6 | 910 | 64 |
AK172848.1 | - | 5 - 6 | 801 | 64 | |
AK314044.1 | - | 5 - 6 | 789 | 64 | |
AL136567.1 | - | 5 - 6 | 685 | 64 | |
BC008920.2 | - | 1 - 2 | 61 | 64 | |
BC010064.2 | - | 5 - 6 | 770 | 64 | |
BC013283.2 | - | 5 - 6 | 1131 | 64 | |
BC019091.2 | - | 5 - 6 | 770 | 64 | |
BC025926.1 | - | 5 - 6 | 766 | 64 | |
CR533566.1 | - | 5 - 6 | 667 | 64 |