Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL20
Entrez Gene ID
27252
Gene Name
kelch like family member 20
Gene Aliases
KHLHX, KLEIP, KLHLX
Chromosome Location
Chr.1: 173714912 - 173786826 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014458.3 NP_055273.2 8 - 9 1477 64
XM_005245093.3 XP_005245150.1 9 - 10 1626 64
XM_017001052.1 XP_016856541.1 8 - 9 1756 64
GenBank mRNA AB026190.1 - 8 - 9 1418 64
AJ844466.1 - 7 - 8 1298 64
AK001430.1 - 8 - 9 1434 64
AK300755.1 - 7 - 8 874 64
AK308048.1 - 7 - 8 1395 64
BC063418.1 - 8 - 9 1407 64

View Full Product Details