The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017873.3 | NP_060343.1 | 5 - 6 | 769 | 87 |
NM_177999.2 | NP_821066.1 | 4 - 5 | 660 | 87 | |
GenBank mRNA | AK000555.1 | - | 5 - 6 | 766 | 87 |
AK023004.1 | - | 5 - 6 | 715 | 87 | |
AK299193.1 | - | 4 - 5 | 564 | 87 | |
BC001719.1 | - | 4 - 5 | 615 | 87 | |
BC065913.1 | - | 5 - 6 | 672 | 87 | |
BX648923.1 | - | 4 - 5 | 1290 | 87 |