The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024619.3 | NP_078895.2 | 3 - 4 | 431 | 69 |
NR_046408.1 | 4 - 5 | 559 | 69 | ||
GenBank mRNA | AK022233.1 | - | 3 - 4 | 398 | 69 |
AK301958.1 | - | 4 - 5 | 543 | 69 | |
AK315373.1 | - | 3 - 4 | 415 | 69 | |
AY360465.1 | - | 3 - 4 | 398 | 69 | |
BC007611.1 | - | 3 - 4 | 407 | 69 | |
BC014408.1 | - | 3 - 4 | 385 | 69 | |
DA019669.1 | - | 3 - 4 | 432 | 69 |