Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX7
Entrez Gene ID
51375
Gene Name
sorting nexin 7
Gene Aliases
-
Chromosome Location
Chr.1: 98661344 - 98760500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015976.4 NP_057060.2 1 - 2 234 69
NM_152238.3 NP_689424.2 1 - 2 234 69
XM_006710677.2 XP_006710740.1 1 - 2 251 69
GenBank mRNA AF121857.1 - 1 - 2 77 69
AK025031.1 - 1 - 2 191 69
AK223229.1 - 1 - 2 93 69
AK298211.1 - 1 - 2 80 69
AL049989.1 - 1 - 2 91 69
AL548028.3 - 1 - 2 235 69
BC010349.1 - 1 - 2 48 69

View Full Product Details