Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX7
Entrez Gene ID
51375
Gene Name
sorting nexin 7
Gene Aliases
-
Chromosome Location
Chr.1: 98661344 - 98760500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015976.4 NP_057060.2 7 - 8 1174 120
NM_152238.3 NP_689424.2 6 - 7 1009 120
XM_006710678.2 XP_006710741.1 8 - 9 1290 120
XM_011541564.2 XP_011539866.1 8 - 9 1230 120
XM_017001425.1 XP_016856914.1 7 - 8 1152 120
XM_017001426.1 XP_016856915.1 8 - 9 1289 120
XM_017001427.1 XP_016856916.1 8 - 9 1564 120
GenBank mRNA AF121857.1 - 7 - 8 1017 120
AK025031.1 - 7 - 8 1131 120
AK223229.1 - 7 - 8 1033 120
AK293018.1 - 7 - 8 1102 120
AK298211.1 - 6 - 7 855 120
BC010349.1 - 7 - 8 988 120

View Full Product Details