The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166347.1 | NP_001159819.1 | 9 - 10 | 1294 | 76 |
NM_001166348.1 | NP_001159820.1 | 8 - 9 | 1224 | 76 | |
NM_001166349.1 | NP_001159821.1 | 8 - 9 | 1094 | 76 | |
NM_173626.3 | NP_775897.3 | 9 - 10 | 1291 | 76 | |
XM_006721833.1 | XP_006721896.1 | 9 - 10 | 1366 | 76 | |
XM_011524652.1 | XP_011522954.1 | 7 - 8 | 1127 | 76 | |
XM_011524653.1 | XP_011522955.1 | 7 - 8 | 1055 | 76 | |
XM_011524654.1 | XP_011522956.1 | 7 - 8 | 940 | 76 | |
XM_017024505.1 | XP_016879994.1 | 8 - 9 | 1280 | 76 | |
XM_017024506.1 | XP_016879995.1 | 8 - 9 | 1280 | 76 | |
XM_017024507.1 | XP_016879996.1 | 9 - 10 | 1340 | 76 | |
GenBank mRNA | AF331524.1 | - | 1 - 2 | 68 | 76 |
AK075248.1 | - | 9 - 10 | 1254 | 76 | |
AK315132.1 | - | 9 - 10 | 1257 | 76 | |
AL833468.1 | - | 8 - 9 | 1188 | 76 | |
BC035900.1 | - | 9 - 10 | 1233 | 76 | |
BC047451.1 | - | 3 - 4 | 343 | 76 |