The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014286.3 | NP_055101.2 | 1 - 2 | 143 | 72 |
GenBank mRNA | AF134479.1 | - | 1 - 2 | 124 | 72 |
AF186409.1 | - | 1 - 2 | 70 | 72 | |
BC004856.2 | - | 1 - 2 | 143 | 72 | |
X84048.1 | - | 1 - 2 | 71 | 72 |