Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
Chromosome Location
Chr.3: 180614008 - 180679495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181426.1 NP_852091.1 12 - 13 1780 107
GenBank mRNA AL122120.1 - 18 - 19 2537 107

View Full Product Details