Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
Chromosome Location
Chr.3: 180614008 - 180679495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181426.1 NP_852091.1 16 - 17 2378 159
GenBank mRNA AK304270.1 - 16 - 17 2357 159
AL122120.1 - 21 - 22 2975 159
BG205877.1 - 3 - 4 389 159

View Full Product Details