Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
Chromosome Location
Chr.3: 180614008 - 180679495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181426.1 NP_852091.1 4 - 5 628 121
GenBank mRNA AK304270.1 - 4 - 5 607 121
AL122120.1 - 10 - 11 1385 121

View Full Product Details