The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160423.1 | NP_001153895.1 | 10 - 11 | 1312 | 91 |
NM_006546.3 | NP_006537.3 | 12 - 13 | 1729 | 91 | |
XM_005256955.3 | XP_005257012.2 | 11 - 12 | 1452 | 91 | |
XM_011524201.2 | XP_011522503.1 | 12 - 13 | 1677 | 91 | |
XM_017024022.1 | XP_016879511.1 | 12 - 13 | 1674 | 91 | |
GenBank mRNA | AF117106.1 | - | 12 - 13 | 1404 | 91 |
AF198254.1 | - | 12 - 13 | 1700 | 91 | |
AK074915.1 | - | 7 - 8 | 732 | 91 | |
AK313229.1 | - | 12 - 13 | 1700 | 91 | |
DQ227344.1 | - | 10 - 11 | 978 | 91 | |
KR861348.1 | - | 11 - 12 | 1351 | 91 |