The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040429.2 | NP_001035519.1 | 3 - 4 | 3685 | 90 |
XM_005266357.2 | XP_005266414.1 | 4 - 5 | 5130 | 90 | |
XM_005266358.2 | XP_005266415.1 | 3 - 4 | 5233 | 90 | |
GenBank mRNA | AF029343.1 | - | 3 - 4 | 3538 | 90 |
AK289980.1 | - | 3 - 4 | 3528 | 90 | |
AK307545.1 | - | 4 - 5 | 1419 | 90 | |
BC028165.1 | - | 3 - 4 | 2819 | 90 |