Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAPK1
Entrez Gene ID
1612
Gene Name
death associated protein kinase 1
Gene Aliases
DAPK
Chromosome Location
Chr.9: 87497228 - 87708634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288729.1 NP_001275658.1 25 - 26 3266 80
NM_001288730.1 NP_001275659.1 25 - 26 3256 80
NM_001288731.1 NP_001275660.1 25 - 26 3239 80
NM_004938.3 NP_004929.2 25 - 26 3591 80
XM_017014360.1 XP_016869849.1 12 - 13 2504 80
GenBank mRNA AB208871.1 - 25 - 26 3194 80
AK295119.1 - 25 - 26 3323 80
BC113660.1 - 24 - 25 3129 80
BC143730.1 - 23 - 24 3079 80
BC143733.1 - 24 - 25 3129 80
BC143759.1 - 23 - 24 2931 80
CR749834.1 - 25 - 26 3239 80
CX758421.1 - 2 - 3 254 80
X76104.1 - 25 - 26 3395 80

View Full Product Details