The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168.2 | NP_001159.2 | 2 - 3 | 338 | 79 |
GenBank mRNA | AB154416.1 | - | 2 - 3 | 217 | 79 |
AF077350.1 | - | 2 - 3 | 241 | 79 | |
AK223428.1 | - | 2 - 3 | 283 | 79 | |
AK301793.1 | - | 2 - 3 | 291 | 79 | |
AK311917.1 | - | 2 - 3 | 242 | 79 | |
BC008718.2 | - | 2 - 3 | 250 | 79 | |
BC034148.1 | - | 2 - 3 | 261 | 79 | |
BC065497.1 | - | 2 - 3 | 231 | 79 | |
CR541740.1 | - | 2 - 3 | 217 | 79 | |
HM625836.1 | - | 2 - 3 | 257 | 79 |