Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
Chromosome Location
Chr.3: 15206226 - 15252916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014296.2 NP_055111.1 13 - 14 1821 64
XM_011533555.2 XP_011531857.1 14 - 15 1720 64
XM_011533556.2 XP_011531858.1 15 - 16 1952 64
XM_011533557.2 XP_011531859.1 14 - 15 1842 64
XM_017006043.1 XP_016861532.1 14 - 15 1904 64
XM_017006044.1 XP_016861533.1 13 - 14 1794 64
XM_017006045.1 XP_016861534.1 14 - 15 1860 64
GenBank mRNA AB028639.1 - 13 - 14 1802 64
AK314253.1 - 13 - 14 1783 64
BC056202.1 - 13 - 14 1799 64
BX502535.1 - 4 - 5 480 64

View Full Product Details