The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014296.2 | NP_055111.1 | 18 - 19 | 2338 | 94 |
XM_011533555.2 | XP_011531857.1 | 19 - 20 | 2237 | 94 | |
XM_011533556.2 | XP_011531858.1 | 20 - 21 | 2469 | 94 | |
XM_017006043.1 | XP_016861532.1 | 19 - 20 | 2421 | 94 | |
GenBank mRNA | AB028639.1 | - | 18 - 19 | 2319 | 94 |
AK314253.1 | - | 18 - 19 | 2300 | 94 | |
BC056202.1 | - | 18 - 19 | 2316 | 94 |