The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003853.3 | NP_003844.1 | 7 - 8 | 1282 | 116 |
XM_011512087.2 | XP_011510389.1 | 7 - 8 | 1056 | 116 | |
XM_011512088.2 | XP_011510390.1 | 4 - 5 | 504 | 116 | |
GenBank mRNA | AF077346.1 | - | 7 - 8 | 1277 | 116 |
BC069630.1 | - | 5 - 6 | 879 | 116 | |
BC106764.2 | - | 5 - 6 | 681 | 116 | |
BC106765.2 | - | 6 - 7 | 1006 | 116 | |
BC137474.1 | - | 5 - 6 | 822 | 116 | |
BC137475.1 | - | 5 - 6 | 822 | 116 |