The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142605.1 | NP_001136077.1 | 20 - 21 | 2288 | 66 |
NM_001258353.1 | NP_001245282.1 | 21 - 22 | 2534 | 66 | |
NM_001258354.1 | NP_001245283.1 | 21 - 22 | 2367 | 66 | |
NM_004247.3 | NP_004238.3 | 21 - 22 | 2397 | 66 | |
GenBank mRNA | AJ505017.1 | - | 17 - 18 | 1771 | 66 |
AK126193.1 | - | 11 - 12 | 1864 | 66 | |
AK126464.1 | - | 22 - 23 | 2203 | 66 | |
AK126652.1 | - | 5 - 6 | 1795 | 66 | |
AK128148.1 | - | 21 - 22 | 2535 | 66 | |
AK225335.1 | - | 21 - 22 | 2157 | 66 | |
AK293108.1 | - | 21 - 22 | 2160 | 66 | |
AK296367.1 | - | 20 - 21 | 2091 | 66 | |
AK297392.1 | - | 21 - 22 | 2173 | 66 | |
AK302828.1 | - | 21 - 22 | 2037 | 66 | |
AK316098.1 | - | 20 - 21 | 2108 | 66 | |
BC002360.2 | - | 21 - 22 | 2167 | 66 | |
BX647305.1 | - | 3 - 4 | 744 | 66 | |
CR456774.1 | - | 20 - 21 | 2136 | 66 | |
D21163.1 | - | 21 - 22 | 2196 | 66 |