The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142605.1 | NP_001136077.1 | 5 - 6 | 648 | 72 |
NM_001258353.1 | NP_001245282.1 | 6 - 7 | 894 | 72 | |
NM_001258354.1 | NP_001245283.1 | 6 - 7 | 727 | 72 | |
NM_004247.3 | NP_004238.3 | 6 - 7 | 757 | 72 | |
GenBank mRNA | AJ505017.1 | - | 2 - 3 | 131 | 72 |
AK126464.1 | - | 7 - 8 | 563 | 72 | |
AK128148.1 | - | 6 - 7 | 894 | 72 | |
AK225335.1 | - | 6 - 7 | 517 | 72 | |
AK297392.1 | - | 6 - 7 | 533 | 72 | |
AK302828.1 | - | 6 - 7 | 397 | 72 | |
AK316098.1 | - | 5 - 6 | 468 | 72 | |
BC002360.2 | - | 6 - 7 | 527 | 72 | |
CR456774.1 | - | 5 - 6 | 496 | 72 | |
D21163.1 | - | 6 - 7 | 556 | 72 |