The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142605.1 | NP_001136077.1 | 7 - 8 | 772 | 85 |
NM_001258353.1 | NP_001245282.1 | 8 - 9 | 1018 | 85 | |
NM_001258354.1 | NP_001245283.1 | 8 - 9 | 851 | 85 | |
NM_004247.3 | NP_004238.3 | 8 - 9 | 881 | 85 | |
GenBank mRNA | AJ505017.1 | - | 4 - 5 | 255 | 85 |
AK126464.1 | - | 9 - 10 | 687 | 85 | |
AK128148.1 | - | 8 - 9 | 1018 | 85 | |
AK225335.1 | - | 8 - 9 | 641 | 85 | |
AK293108.1 | - | 8 - 9 | 644 | 85 | |
AK296367.1 | - | 7 - 8 | 575 | 85 | |
AK297392.1 | - | 8 - 9 | 657 | 85 | |
AK302828.1 | - | 8 - 9 | 521 | 85 | |
AK316098.1 | - | 7 - 8 | 592 | 85 | |
BC002360.2 | - | 8 - 9 | 651 | 85 | |
CR456774.1 | - | 7 - 8 | 620 | 85 | |
D21163.1 | - | 8 - 9 | 680 | 85 |