The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317926.1 | NP_001304855.1 | 13 - 14 | 2064 | 69 |
NM_013355.4 | NP_037487.2 | 13 - 14 | 2064 | 69 | |
XM_005251946.3 | XP_005252003.1 | 13 - 14 | 2619 | 69 | |
XM_006717080.2 | XP_006717143.1 | 13 - 14 | 2133 | 69 | |
XM_017014649.1 | XP_016870138.1 | 13 - 14 | 2135 | 69 | |
XM_017014650.1 | XP_016870139.1 | 13 - 14 | 2273 | 69 | |
GenBank mRNA | AB019692.1 | - | 13 - 14 | 1652 | 69 |
BC062558.1 | - | 13 - 14 | 1667 | 69 |