The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320126.1 | NP_001307055.1 | 9 - 10 | 1341 | 80 |
NM_020676.6 | NP_065727.4 | 8 - 9 | 1276 | 80 | |
GenBank mRNA | AK122983.1 | - | 9 - 10 | 1033 | 80 |
AK172797.1 | - | 8 - 9 | 1197 | 80 | |
AK225942.1 | - | 10 - 11 | 1550 | 80 | |
AK313168.1 | - | 8 - 9 | 1167 | 80 | |
BC001698.2 | - | 8 - 9 | 949 | 80 |