Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNH1
Entrez Gene ID
6050
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
RAI, RNH
Chromosome Location
Chr.11: 494512 - 507283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002939.3 NP_002930.2 6 - 7 1082 79
NM_203383.1 NP_976317.1 6 - 7 1014 79
NM_203384.1 NP_976318.1 5 - 6 841 79
NM_203385.1 NP_976319.1 5 - 6 909 79
NM_203386.2 NP_976320.1 6 - 7 972 79
NM_203387.2 NP_976321.1 6 - 7 1040 79
NM_203388.2 NP_976322.1 5 - 6 799 79
NM_203389.2 NP_976323.1 5 - 6 867 79
XM_011520255.1 XP_011518557.1 6 - 7 1017 79
XM_011520257.2 XP_011518559.1 6 - 7 1265 79
XM_011520258.2 XP_011518560.1 6 - 7 1233 79
XM_011520259.2 XP_011518561.1 5 - 6 782 79
XM_011520260.2 XP_011518562.1 6 - 7 941 79
XM_011520261.2 XP_011518563.1 6 - 7 1015 79
XM_011520262.2 XP_011518564.1 6 - 7 1198 79
XM_011520263.1 XP_011518565.1 5 - 6 749 79
XM_017018106.1 XP_016873595.1 6 - 7 1082 79
GenBank mRNA AB209196.1 - 4 - 5 2332 79
AL161967.1 - 4 - 5 2017 79
BC000677.2 - 5 - 6 769 79
BC003506.1 - 5 - 6 782 79
BC011186.1 - 4 - 5 567 79
BC011500.1 - 5 - 6 720 79
BC014629.2 - 5 - 6 769 79
BC024037.1 - 5 - 6 782 79
BC047730.1 - 6 - 7 1061 79
M22414.1 - 5 - 6 730 79
M36717.1 - 6 - 7 969 79
X13973.1 - 6 - 7 969 79

View Full Product Details