Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNH1
Entrez Gene ID
6050
Gene Name
ribonuclease/angiogenin inhibitor 1
Gene Aliases
RAI, RNH
Chromosome Location
Chr.11: 494512 - 507283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002939.3 NP_002930.2 8 - 9 1428 71
NM_203383.1 NP_976317.1 8 - 9 1360 71
NM_203384.1 NP_976318.1 7 - 8 1187 71
NM_203385.1 NP_976319.1 7 - 8 1255 71
NM_203386.2 NP_976320.1 8 - 9 1318 71
NM_203387.2 NP_976321.1 8 - 9 1386 71
NM_203388.2 NP_976322.1 7 - 8 1145 71
NM_203389.2 NP_976323.1 7 - 8 1213 71
XM_011520255.1 XP_011518557.1 8 - 9 1363 71
XM_011520257.2 XP_011518559.1 8 - 9 1611 71
XM_011520258.2 XP_011518560.1 8 - 9 1579 71
XM_011520259.2 XP_011518561.1 7 - 8 1128 71
XM_011520260.2 XP_011518562.1 8 - 9 1287 71
XM_011520261.2 XP_011518563.1 8 - 9 1361 71
XM_011520262.2 XP_011518564.1 8 - 9 1544 71
XM_011520263.1 XP_011518565.1 7 - 8 1095 71
XM_017018106.1 XP_016873595.1 8 - 9 1428 71
GenBank mRNA AB209196.1 - 6 - 7 2678 71
AL161967.1 - 6 - 7 2363 71
BC000677.2 - 7 - 8 1115 71
BC003506.1 - 7 - 8 1128 71
BC011186.1 - 6 - 7 913 71
BC011500.1 - 7 - 8 1066 71
BC014629.2 - 7 - 8 1115 71
BC024037.1 - 7 - 8 1128 71
BC047730.1 - 8 - 9 1407 71
M22414.1 - 7 - 8 1076 71
M36717.1 - 8 - 9 1315 71
X13973.1 - 8 - 9 1315 71

View Full Product Details