Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
Chromosome Location
Chr.17: 73193034 - 73208507 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018714.2 NP_061184.1 2 - 3 598 100
GenBank mRNA AB037802.1 - 2 - 3 560 100
AK098224.1 - 2 - 3 318 100
BC021985.2 - 2 - 3 563 100
BC047465.1 - 3 - 4 900 100

View Full Product Details