Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFASC
Entrez Gene ID
23114
Gene Name
neurofascin
Gene Aliases
NF, NRCAML
Chromosome Location
Chr.1: 204828652 - 205022822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005388.2 NP_001005388.2 12 - 13 1600 102
NM_001005389.1 NP_001005389.2 12 - 13 1600 102
NM_001160331.1 NP_001153803.1 10 - 11 1395 102
NM_001160332.1 NP_001153804.1 12 - 13 1633 102
NM_001160333.1 NP_001153805.1 11 - 12 1582 102
NM_015090.3 NP_055905.2 12 - 13 1633 102
XM_005244989.3 XP_005245046.2 12 - 13 1713 102
XM_005244991.3 XP_005245048.2 12 - 13 1713 102
XM_005244992.4 XP_005245049.2 12 - 13 1713 102
XM_005244993.3 XP_005245050.2 11 - 12 1662 102
XM_005244997.3 XP_005245054.2 11 - 12 1662 102
XM_011509311.1 XP_011507613.1 11 - 12 1413 102
XM_011509312.1 XP_011507614.1 12 - 13 1713 102
XM_011509313.1 XP_011507615.1 11 - 12 1413 102
XM_011509314.1 XP_011507616.1 11 - 12 1662 102
XM_011509315.1 XP_011507617.1 10 - 11 1362 102
XM_011509316.1 XP_011507618.1 13 - 14 1610 102
XM_011509317.1 XP_011507619.1 11 - 12 1413 102
XM_011509318.2 XP_011507620.1 12 - 13 1635 102
XM_011509319.1 XP_011507621.1 12 - 13 1559 102
XM_011509320.2 XP_011507622.1 11 - 12 1584 102
XM_011509321.1 XP_011507623.1 11 - 12 1413 102
XM_011509322.1 XP_011507624.1 11 - 12 1413 102
XM_011509323.1 XP_011507625.1 11 - 12 1413 102
XM_011509324.1 XP_011507626.1 11 - 12 1413 102
XM_011509325.1 XP_011507627.1 11 - 12 1413 102
XM_011509326.1 XP_011507628.1 11 - 12 1413 102
XM_011509327.1 XP_011507629.1 11 - 12 1413 102
XM_011509328.1 XP_011507630.1 10 - 11 1362 102
XM_017000733.1 XP_016856222.1 12 - 13 1713 102
XM_017000734.1 XP_016856223.1 11 - 12 1662 102
XM_017000735.1 XP_016856224.1 11 - 12 1413 102
XM_017000736.1 XP_016856225.1 11 - 12 1413 102
XM_017000737.1 XP_016856226.1 11 - 12 1413 102
XM_017000738.1 XP_016856227.1 12 - 13 1635 102
XM_017000739.1 XP_016856228.1 12 - 13 1635 102
XM_017000740.1 XP_016856229.1 12 - 13 1635 102
XM_017000741.1 XP_016856230.1 9 - 10 1203 102
XM_017000742.1 XP_016856231.1 10 - 11 1349 102
XM_017000743.1 XP_016856232.1 11 - 12 1584 102
GenBank mRNA AB018299.3 - 11 - 12 1464 102
AB177861.1 - 5 - 6 698 102
AK090639.1 - 12 - 13 1559 102
AK295291.1 - 11 - 12 1561 102
AK299262.1 - 11 - 12 1561 102
AK308686.1 - 12 - 13 1713 102
BC008124.2 - 3 - 4 298 102
BC040674.1 - 13 - 14 1851 102
BC117674.2 - 11 - 12 1449 102
BC137013.1 - 10 - 11 1370 102
BC144454.1 - 10 - 11 1337 102
BC144455.1 - 11 - 12 1388 102

View Full Product Details