The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000342.3 | NP_000333.1 | 10 - 11 | 1231 | 73 |
XM_005257593.4 | XP_005257650.1 | 8 - 9 | 999 | 73 | |
XM_011525129.1 | XP_011523431.1 | 10 - 11 | 1258 | 73 | |
XM_011525130.1 | XP_011523432.1 | 10 - 11 | 1258 | 73 | |
GenBank mRNA | AB526447.1 | - | 8 - 9 | 1082 | 73 |
BC096106.3 | - | 9 - 10 | 1144 | 73 | |
BC096107.3 | - | 9 - 10 | 1144 | 73 | |
BC099628.3 | - | 9 - 10 | 1144 | 73 | |
BC099629.3 | - | 9 - 10 | 1144 | 73 | |
BC101570.1 | - | 10 - 11 | 1168 | 73 | |
BC101574.1 | - | 9 - 10 | 1164 | 73 | |
DQ419529.1 | - | 9 - 10 | 1150 | 73 | |
GQ981383.1 | - | 9 - 10 | 1125 | 73 | |
GQ981384.1 | - | 9 - 10 | 1125 | 73 | |
GQ981386.1 | - | 9 - 10 | 1125 | 73 | |
GQ981387.1 | - | 9 - 10 | 1125 | 73 | |
M27819.1 | - | 10 - 11 | 1196 | 73 | |
X12609.1 | - | 10 - 11 | 1232 | 73 |