Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PITPNB
Entrez Gene ID
23760
Gene Name
phosphatidylinositol transfer protein beta
Gene Aliases
PI-TP-beta, PtdInsTP, VIB1B
Chromosome Location
Chr.22: 27851669 - 27919306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284277.1 NP_001271206.1 8 - 9 650 89
NM_001284278.1 NP_001271207.1 8 - 9 628 89
NM_012399.4 NP_036531.1 8 - 9 650 89
XM_011530052.2 XP_011528354.1 8 - 9 663 89
GenBank mRNA AK123384.1 - 1 - 2 84 89
AK302367.1 - 8 - 9 614 89
AK313064.1 - 8 - 9 600 89
BC018704.1 - 8 - 9 574 89
BC031427.1 - 8 - 9 567 89
BU147254.1 - 8 - 9 611 89
CR456541.1 - 8 - 9 602 89
D30037.1 - 8 - 9 581 89

View Full Product Details