The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163.3 | NP_001154.2 | 1 - 2 | 205 | 91 |
XM_005251968.3 | XP_005252025.1 | 1 - 2 | 236 | 91 | |
GenBank mRNA | AF029106.1 | - | 1 - 2 | 127 | 91 |
AF047347.1 | - | 1 - 2 | 168 | 91 | |
BQ269294.1 | - | 1 - 2 | 213 | 91 |