The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033058.2 | NP_149047.2 | 6 - 7 | 1081 | 124 |
NM_184085.1 | NP_908973.1 | 6 - 7 | 1081 | 124 | |
NM_184086.1 | NP_908974.1 | 6 - 7 | 1081 | 124 | |
XM_005251316.4 | XP_005251373.1 | 8 - 9 | 2106 | 124 | |
XM_011517613.2 | XP_011515915.1 | 8 - 9 | 2106 | 124 | |
XM_011517614.2 | XP_011515916.1 | 8 - 9 | 2029 | 124 | |
XM_011517615.2 | XP_011515917.1 | 8 - 9 | 2106 | 124 | |
XM_011517616.2 | XP_011515918.1 | 8 - 9 | 2106 | 124 | |
XM_011517617.2 | XP_011515919.1 | 6 - 7 | 1163 | 124 | |
XM_017013908.1 | XP_016869397.1 | 6 - 7 | 799 | 124 | |
XM_017013909.1 | XP_016869398.1 | 5 - 6 | 618 | 124 | |
GenBank mRNA | AJ243488.1 | - | 6 - 7 | 1081 | 124 |
AJ243489.1 | - | 6 - 7 | 1081 | 124 | |
AJ291712.1 | - | 6 - 7 | 987 | 124 | |
AJ431704.1 | - | 6 - 7 | 1081 | 124 | |
AK091310.1 | - | 6 - 7 | 1231 | 124 | |
AK091728.1 | - | 6 - 7 | 901 | 124 | |
BC007750.2 | - | 6 - 7 | 987 | 124 | |
BT007212.1 | - | 6 - 7 | 807 | 124 |