The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000042.2 | NP_000033.2 | 3 - 4 | 393 | 138 |
GenBank mRNA | AK298416.1 | - | 3 - 4 | 365 | 138 |
AK313838.1 | - | 3 - 4 | 349 | 138 | |
BC020703.1 | - | 3 - 4 | 351 | 138 | |
BC026283.1 | - | 3 - 4 | 365 | 138 | |
BG566874.1 | - | 3 - 4 | 393 | 138 | |
HM212425.1 | - | 2 - 3 | 277 | 138 | |
M62839.1 | - | 3 - 4 | 373 | 138 | |
S80305.1 | - | 3 - 4 | 343 | 138 | |
X53595.1 | - | 3 - 4 | 367 | 138 | |
X57847.1 | - | 3 - 4 | 367 | 138 | |
X58100.1 | - | 3 - 4 | 355 | 138 |