Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL36A
Entrez Gene ID
6173
Gene Name
ribosomal protein L36a
Gene Aliases
L36A, L44L, MIG6, RPL44
Chromosome Location
Chr.X: 101390890 - 101396154 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199972.1 NP_001186901.1 1 - 2 153 59
NM_021029.5 NP_066357.2 1 - 2 153 59
GenBank mRNA AK096711.1 - 1 - 2 32 59
BC001781.1 - 1 - 1 39 59
BC031015.1 - 1 - 1 31 59
BC062219.1 - 1 - 2 64 59
BC070204.1 - 1 - 1 28 59
BF211310.1 - 1 - 2 84 59
BG504049.1 - 1 - 2 32 59
BG943625.1 - 1 - 2 220 59
BP245950.1 - 1 - 2 153 59

Gene Symbol
RPL36A-HNRNPH2
Entrez Gene ID
100529097
Gene Name
RPL36A-HNRNPH2 readthrough
Gene Aliases
-
Chromosome Location
Chr.X: 101390890 - 101414140 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199973.1 NP_001186902.1 1 - 2 153 59
NM_001199974.1 NP_001186903.1 1 - 2 153 59
GenBank mRNA DA530036.1 - 1 - 2 28 59
DA998453.1 - 1 - 2 32 59

View Full Product Details