The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000350.2 | NP_000341.2 | 33 - 34 | 4879 | 133 |
GenBank mRNA | AB210040.1 | - | 33 - 34 | 4865 | 133 |
AF000148.1 | - | 33 - 34 | 4862 | 133 | |
AF001945.1 | - | 33 - 34 | 4856 | 133 | |
CR627391.1 | - | 11 - 12 | 1291 | 133 | |
U66691.1 | - | 10 - 11 | 1160 | 133 | |
U88667.1 | - | 33 - 34 | 4856 | 133 |